siRNA Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162411.1 | NP_001155883.1 | 5 | 1061 | ||
| XM_008767249.1 | XP_008765471.1 | 5 | 1084 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha4| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162411.1 | NP_001155883.1 | 5 | 1061 | ||
| XM_008767249.1 | XP_008765471.1 | 5 | 1084 | |||