siRNA Details
| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108228.1 | NP_001101698.1 | 4 | 1044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc19a3| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108228.1 | NP_001101698.1 | 4 | 1044 | ||