siRNA Details
| Gene Symbol: Slc16a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108229.1 | NP_001101699.1 | 3 | 749 | ||
| XM_006245388.2 | XP_006245450.1 | 3 | 978 | |||
| XM_006245389.2 | XP_006245451.1 | 3 | 1125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a14| Gene Symbol: Slc16a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108229.1 | NP_001101699.1 | 3 | 749 | ||
| XM_006245388.2 | XP_006245450.1 | 3 | 978 | |||
| XM_006245389.2 | XP_006245451.1 | 3 | 1125 | |||