siRNA Details
| Gene Symbol: Mlph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012135.1 | NP_001012135.1 | 10 | 1334 | ||
| XM_006245404.2 | XP_006245466.1 | 10 | 1535 | |||
| GenBank mRNA | BC081894.1 | - | 10 | 1334 | 7110013 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlph| Gene Symbol: Mlph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012135.1 | NP_001012135.1 | 10 | 1334 | ||
| XM_006245404.2 | XP_006245466.1 | 10 | 1535 | |||
| GenBank mRNA | BC081894.1 | - | 10 | 1334 | 7110013 | |