siRNA Details
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750733.2 | XP_003750781.1 | 3 | 1054 | ||
GenBank mRNA | FQ216933.1 | - | 3 | 1022 | ||
FQ216966.1 | - | 3 | 1054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750733.2 | XP_003750781.1 | 3 | 1054 | ||
GenBank mRNA | FQ216933.1 | - | 3 | 1022 | ||
FQ216966.1 | - | 3 | 1054 |