siRNA Details
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025711.1 | NP_001020882.1 | 3, 4 | 399 | ||
XM_006245533.2 | XP_006245595.1 | 9, 10 | 939 | |||
XM_006245534.2 | XP_006245596.1 | 8, 9 | 842 | |||
GenBank mRNA | BC098833.1 | - | 3, 4 | 399 | 7378703 |
