siRNA Details
| Gene Symbol: Myom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191584.1 | NP_001178513.1 | 26 | 3905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myom1| Gene Symbol: Myom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191584.1 | NP_001178513.1 | 26 | 3905 | ||