siRNA Details
| Gene Symbol: Lama1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108237.2 | NP_001101707.2 | 54, 55 | 8288 | ||
| GenBank mRNA | AB425340.1 | - | 54, 55 | 8288 | ||
| AB425341.1 | - | 54, 55 | 8288 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Lama1
Lama1| Gene Symbol: Lama1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108237.2 | NP_001101707.2 | 54, 55 | 8288 | ||
| GenBank mRNA | AB425340.1 | - | 54, 55 | 8288 | ||
| AB425341.1 | - | 54, 55 | 8288 | |||
 Back To Top
 Back To Top