siRNA Details
| Gene Symbol: Lama1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108237.2 | NP_001101707.2 | 43, 44 | 6717 | ||
| GenBank mRNA | AB425340.1 | - | 43, 44 | 6717 | ||
| AB425341.1 | - | 43, 44 | 6717 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lama1| Gene Symbol: Lama1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108237.2 | NP_001101707.2 | 43, 44 | 6717 | ||
| GenBank mRNA | AB425340.1 | - | 43, 44 | 6717 | ||
| AB425341.1 | - | 43, 44 | 6717 | |||