siRNA Details
| Gene Symbol: Ankrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108238.1 | NP_001101708.1 | 8 | 5031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd12| Gene Symbol: Ankrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108238.1 | NP_001101708.1 | 8 | 5031 | ||