siRNA Details
| Gene Symbol: Ankrd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108238.1 | NP_001101708.1 | 2 | 277 | ||
| Gene Symbol: LOC100910483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767402.1 | XP_008765624.1 | 2 | 2903 | ||
| XM_008767403.1 | XP_008765625.1 | 3 | 403 | |||
| XM_008767404.1 | XP_008765626.1 | 3 | 391 | |||
| XM_008767405.1 | XP_008765627.1 | 2 | 2903 | |||

Ankrd12
Genomic Map