siRNA Details
| Gene Symbol: Clptm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108240.1 | NP_001101710.1 | 8 | 1156 | ||
| XM_006227791.2 | XP_006227853.1 | 8 | 1158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clptm1l| Gene Symbol: Clptm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108240.1 | NP_001101710.1 | 8 | 1156 | ||
| XM_006227791.2 | XP_006227853.1 | 8 | 1158 | |||