siRNA Details
| Gene Symbol: Magee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079891.1 | NP_001073360.1 | 1 | 2034 | ||
| GenBank mRNA | BC128754.1 | - | 1 | 2034 | 7103930 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magee1| Gene Symbol: Magee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079891.1 | NP_001073360.1 | 1 | 2034 | ||
| GenBank mRNA | BC128754.1 | - | 1 | 2034 | 7103930 | |