siRNA Details
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039101.1 | NP_001034190.1 | 9 | 1036 | ||
XM_006257145.2 | XP_006257207.1 | 9 | 1093 | |||
XM_008773347.1 | XP_008771569.1 | 10 | 1856 | |||
XM_008773348.1 | XP_008771570.1 | 11 | 1009 | |||
GenBank mRNA | AY886531.1 | - | 9 | 1036 |
