siRNA Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108245.2 | NP_001101715.1 | 11 | 924 | ||
| GenBank mRNA | FQ215476.1 | - | 11 | 919 | ||
| FQ216044.1 | - | 11 | 923 | |||
| FQ216100.1 | - | 11 | 900 | |||
| FQ216185.1 | - | 11 | 921 | |||

Itgb1bp2
Genomic Map