siRNA Details
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012356.1 | NP_001012356.1 | 6 | 853 | ||
GenBank mRNA | BC089880.1 | - | 6 | 853 | 7366190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012356.1 | NP_001012356.1 | 6 | 853 | ||
GenBank mRNA | BC089880.1 | - | 6 | 853 | 7366190 |