siRNA Details
| Gene Symbol: Zmym3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040155.2 | NP_001035245.2 | 1 | 604 | ||
| GenBank mRNA | BC127542.1 | - | 1 | 556 | ||
| FQ228841.1 | - | 1 | 520 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmym3| Gene Symbol: Zmym3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040155.2 | NP_001035245.2 | 1 | 604 | ||
| GenBank mRNA | BC127542.1 | - | 1 | 556 | ||
| FQ228841.1 | - | 1 | 520 | |||