siRNA Details
| Gene Symbol: Bcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191586.1 | NP_001178515.1 | 4 | 2337 | ||
| XM_006256659.2 | XP_006256721.1 | 4 | 2297 | |||
| XM_006256660.2 | XP_006256722.1 | 4 | 2367 | |||
| XM_006256661.2 | XP_006256723.1 | 4 | 2461 | |||
| XM_006256663.2 | XP_006256725.1 | 4 | 2405 | |||
| XM_006256664.2 | XP_006256726.1 | 4 | 2405 | |||
| XM_006256665.2 | XP_006256727.1 | 4 | 2405 | |||
| XM_008773074.1 | XP_008771296.1 | 4 | 7905 | |||

Bcor
Genomic Map