siRNA Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014110.1 | NP_001014132.1 | 4 | 447 | ||
| GenBank mRNA | BC085918.1 | - | 4 | 447 | 7108577 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prickle3| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014110.1 | NP_001014132.1 | 4 | 447 | ||
| GenBank mRNA | BC085918.1 | - | 4 | 447 | 7108577 | |