siRNA Details
Gene Symbol: Usp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108252.1 | NP_001101722.1 | 2 | 901 | ||
XM_006256768.2 | XP_006256830.1 | 4 | 1233 | |||
XM_008773076.1 | XP_008771298.1 | 3 | 1971 |
