siRNA Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207611.1 | NP_997494.1 | 1 | 315 | ||
| XM_006257273.2 | XP_006257335.1 | 4 | 3121 | |||
| XM_006257274.2 | XP_006257336.1 | 5 | 2972 | |||
| XM_006257275.2 | XP_006257337.1 | 4 | 1935 | |||
| XM_006257276.2 | XP_006257338.1 | 5 | 2083 | |||
| XM_008773421.1 | XP_008771643.1 | 4 | 1917 | |||
| GenBank mRNA | AF497483.1 | - | 1 | 315 | ||

Bhlhb9
Genomic Map