siRNA Details
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108253.1 | NP_001101723.1 | 16, 17 | 2227 | ||
XM_006256792.2 | XP_006256854.1 | 16, 17 | 3059 | |||
XM_006256794.2 | XP_006256856.1 | 16, 17 | 2467 | |||
XM_006256795.2 | XP_006256857.1 | 15, 16 | 2756 | |||
XM_006256796.2 | XP_006256858.1 | 16, 17 | 3058 | |||
XM_008773119.1 | XP_008771341.1 | 16, 17 | 2197 |
