siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047893.3 | NP_001041358.2 | 4 | 775 | ||
XM_006256816.2 | XP_006256878.1 | 4 | 872 | |||
XM_006256817.2 | XP_006256879.1 | 4 | 872 | |||
XM_008773126.1 | XP_008771348.1 | 3 | 740 | |||
GenBank mRNA | AY325213.1 | - | 2 | 376 |
