siRNA Details
| Gene Symbol: Ocrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108256.2 | NP_001101726.2 | 11 | 1225 | ||
| XM_006257528.1 | XP_006257590.1 | 10 | 984 | |||
| XM_006257529.1 | XP_006257591.1 | 10 | 984 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ocrl| Gene Symbol: Ocrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108256.2 | NP_001101726.2 | 11 | 1225 | ||
| XM_006257528.1 | XP_006257590.1 | 10 | 984 | |||
| XM_006257529.1 | XP_006257591.1 | 10 | 984 | |||