siRNA Details
| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191736.1 | NP_001178665.1 | 8 | 1117 | ||
| XM_006257573.2 | XP_006257635.1 | 10 | 1604 | |||
| XM_008773567.1 | XP_008771789.1 | 9 | 2316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk26| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191736.1 | NP_001178665.1 | 8 | 1117 | ||
| XM_006257573.2 | XP_006257635.1 | 10 | 1604 | |||
| XM_008773567.1 | XP_008771789.1 | 9 | 2316 | |||