siRNA Details
| Gene Symbol: Htatsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108259.1 | NP_001101729.1 | 10 | 2221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htatsf1| Gene Symbol: Htatsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108259.1 | NP_001101729.1 | 10 | 2221 | ||