siRNA Details
Gene Symbol: Baz2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108260.1 | NP_001101730.1 | 31 | 5480 | ||
XM_008761847.1 | XP_008760069.1 | 29 | 5038 | |||
XM_008761848.1 | XP_008760070.1 | 28 | 4996 | |||
XM_008761849.1 | XP_008760071.1 | 29 | 5032 | |||
XM_008761850.1 | XP_008760072.1 | 28 | 5005 | |||
XM_008761851.1 | XP_008760073.1 | 29 | 5038 | |||
XM_008761852.1 | XP_008760074.1 | 29 | 5026 | |||
XM_008761853.1 | XP_008760075.1 | 28 | 5005 | |||
XM_008761854.1 | XP_008760076.1 | 28 | 4999 | |||
XM_008761855.1 | XP_008760077.1 | 28 | 4849 | |||
XM_008761856.1 | XP_008760078.1 | 27 | 4807 | |||
XM_008761857.1 | XP_008760079.1 | 28 | 4843 | |||
XM_008761858.1 | XP_008760080.1 | 28 | 4750 |
