siRNA Details
| Gene Symbol: Tph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173839.2 | NP_776211.1 | 11 | 1418 | ||
| XM_006241352.2 | XP_006241414.1 | 11 | 1474 | |||
| XM_008765357.1 | XP_008763579.1 | 11 | 1480 | |||
| GenBank mRNA | AY098915.1 | - | 11 | 1418 | ||

Tph2
Genomic Map