siRNA Details
| Gene Symbol: Naglt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176080.2 | NP_788269.1 | 4 | 729 | ||
| GenBank mRNA | AB089802.1 | - | 4 | 729 | ||
| BC098651.1 | - | 4 | 716 | 7130354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naglt1| Gene Symbol: Naglt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176080.2 | NP_788269.1 | 4 | 729 | ||
| GenBank mRNA | AB089802.1 | - | 4 | 729 | ||
| BC098651.1 | - | 4 | 716 | 7130354 | ||