siRNA Details
| Gene Symbol: Sla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178097.2 | NP_835198.1 | 275 | |||
| GenBank mRNA | AY217759.1 | - | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sla| Gene Symbol: Sla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178097.2 | NP_835198.1 | 275 | |||
| GenBank mRNA | AY217759.1 | - | 275 | |||