siRNA Details
| Gene Symbol: Sgms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181386.2 | NP_852051.2 | 8, 9 | 1694 | ||
| GenBank mRNA | AF461433.1 | - | 3 | 379 | ||
| AY280961.1 | - | 4, 5 | 1277 | |||
| BC078690.1 | - | 4, 5 | 1039 | 7113038 | ||
| BC081693.1 | - | 8, 9 | 1694 | 7132545 | ||

Sgms1
Genomic Map