siRNA Details
| Gene Symbol: Spg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181388.2 | NP_852053.1 | 6, 7 | 900 | ||
| XM_008772619.1 | XP_008770841.1 | 6, 7 | 907 | |||
| XM_008772620.1 | XP_008770842.1 | 6, 7 | 881 | |||
| XM_008772621.1 | XP_008770843.1 | 5, 6 | 651 | |||
| GenBank mRNA | AY278739.1 | - | 6, 7 | 900 | ||
| FQ224821.1 | - | 6, 7 | 912 | |||

Spg7
Genomic Map