siRNA Details
| Gene Symbol: Prss21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181477.2 | NP_852142.1 | 4 | 383 | ||
| XM_006245872.2 | XP_006245934.1 | 4 | 801 | |||
| GenBank mRNA | AB074516.1 | - | 4 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss21| Gene Symbol: Prss21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181477.2 | NP_852142.1 | 4 | 383 | ||
| XM_006245872.2 | XP_006245934.1 | 4 | 801 | |||
| GenBank mRNA | AB074516.1 | - | 4 | 383 | ||