siRNA Details
| Gene Symbol: Nadsyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181480.1 | NP_852145.1 | 21 | 2134 | ||
| XM_008760144.1 | XP_008758366.1 | 20 | 2161 | |||
| XM_008760145.1 | XP_008758367.1 | 19 | 1907 | |||
| GenBank mRNA | AB090805.1 | - | 21 | 2134 | ||
| BC161832.1 | - | 21 | 2208 | 9092240 | ||

Nadsyn1
Genomic Map