siRNA Details
| Gene Symbol: Nadsyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181480.1 | NP_852145.1 | 10, 11 | 879 | ||
| XM_008760144.1 | XP_008758366.1 | 9, 10 | 906 | |||
| XM_008760145.1 | XP_008758367.1 | 8, 9 | 652 | |||
| GenBank mRNA | AB090805.1 | - | 10, 11 | 879 | ||
| BC161832.1 | - | 10, 11 | 953 | 9092240 | ||

Nadsyn1
Genomic Map