siRNA Details
Gene Symbol: Gtf2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001512.2 | NP_001001512.2 | 11 | 1169 | ||
XM_006249162.2 | XP_006249224.1 | 15 | 1434 | |||
XM_006249163.2 | XP_006249225.1 | 15 | 1431 | |||
XM_006249164.2 | XP_006249226.1 | 14 | 1374 | |||
XM_006249165.2 | XP_006249227.1 | 14 | 1371 | |||
XM_006249166.2 | XP_006249228.1 | 14 | 1368 | |||
XM_006249167.2 | XP_006249229.1 | 13 | 1317 | |||
XM_006249168.2 | XP_006249230.1 | 13 | 1311 | |||
XM_006249169.2 | XP_006249231.1 | 13 | 1296 | |||
XM_006249170.2 | XP_006249232.1 | 15 | 1629 | |||
XM_006249171.2 | XP_006249233.1 | 12 | 1254 | |||
XM_006249172.2 | XP_006249234.1 | 12 | 1239 | |||
XM_006249173.2 | XP_006249235.1 | 12 | 1233 | |||
XM_006249174.2 | XP_006249236.1 | 11 | 1176 | |||
XM_008769141.1 | XP_008767363.1 | 15 | 1296 | |||
GenBank mRNA | AF547390.1 | - | 13 | 1206 | ||
AF547391.1 | - | 13 | 1206 | |||
BC085815.1 | - | 11 | 1307 | 7123295 |
