siRNA Details
Gene Symbol: Gtf2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001512.2 | NP_001001512.2 | 30 | 2863 | ||
XM_006249162.2 | XP_006249224.1 | 34 | 3128 | |||
XM_006249163.2 | XP_006249225.1 | 34 | 3125 | |||
XM_006249164.2 | XP_006249226.1 | 33 | 3068 | |||
XM_006249165.2 | XP_006249227.1 | 33 | 3065 | |||
XM_006249166.2 | XP_006249228.1 | 33 | 3062 | |||
XM_006249167.2 | XP_006249229.1 | 32 | 3011 | |||
XM_006249168.2 | XP_006249230.1 | 32 | 3005 | |||
XM_006249169.2 | XP_006249231.1 | 32 | 2990 | |||
XM_006249170.2 | XP_006249232.1 | 34 | 3323 | |||
XM_006249171.2 | XP_006249233.1 | 31 | 2948 | |||
XM_006249172.2 | XP_006249234.1 | 31 | 2933 | |||
XM_006249173.2 | XP_006249235.1 | 31 | 2927 | |||
XM_006249174.2 | XP_006249236.1 | 30 | 2870 | |||
XM_008769141.1 | XP_008767363.1 | 34 | 2990 | |||
GenBank mRNA | AF547390.1 | - | 32 | 2900 | ||
AF547391.1 | - | 32 | 2900 | |||
BC085815.1 | - | 30 | 3001 | 7123295 |
