siRNA Details
| Gene Symbol: Pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181637.1 | NP_853668.1 | 10, 11 | 1051 | ||
| XM_006235325.2 | XP_006235387.1 | 11, 12 | 1302 | |||
| XM_008762347.1 | XP_008760569.1 | 8, 9 | 1399 | |||
| GenBank mRNA | AB086841.1 | - | 10, 11 | 1086 | ||
| BC097269.1 | - | 11, 12 | 1177 | |||

Pigu
Genomic Map