siRNA Details
| Gene Symbol: Acsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181695.1 | NP_859046.1 | 9 | 1286 | ||
| GenBank mRNA | AB096688.1 | - | 9 | 1286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsm4| Gene Symbol: Acsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181695.1 | NP_859046.1 | 9 | 1286 | ||
| GenBank mRNA | AB096688.1 | - | 9 | 1286 | ||