siRNA Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182673.2 | NP_872614.1 | 1 | 1291 | ||
| XM_006249998.2 | XP_006250060.1 | 3 | 1678 | |||
| XM_006249999.2 | XP_006250061.1 | 3 | 1882 | |||
| XM_006250000.2 | XP_006250062.1 | 3 | 1968 | |||
| XM_006250001.2 | XP_006250063.1 | 3 | 1657 | |||
| XM_006250002.2 | XP_006250064.1 | 2 | 1733 | |||
| XM_006250004.2 | XP_006250066.1 | 2 | 2245 | |||
| XM_008769653.1 | XP_008767875.1 | 2 | 2245 | |||
| GenBank mRNA | AY262823.1 | - | 1 | 1293 | ||

Rnasel
Genomic Map