siRNA Details
| Gene Symbol: Ston1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182954.2 | NP_891999.2 | 2 | 1326 | ||
| XM_006239743.2 | XP_006239805.1 | 2 | 1097 | |||
| XM_006239744.2 | XP_006239806.1 | 2 | 2193 | |||
| XM_008764479.1 | XP_008762701.1 | 3 | 1414 | |||
| XM_008764480.1 | XP_008762702.1 | 2 | 1882 | |||
| XM_008764481.1 | XP_008762703.1 | 2 | 1077 | |||
| GenBank mRNA | AY300793.1 | - | 1 | 1044 | ||

Ston1
Genomic Map