siRNA Details
| Gene Symbol: Zfp191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182955.1 | NP_892000.1 | 2 | 259 | ||
| XM_006254464.2 | XP_006254526.1 | 2 | 296 | |||
| XM_006254465.2 | XP_006254527.1 | 2 | 654 | |||
| GenBank mRNA | AY336973.1 | - | 2 | 259 | ||

Zfp191
Genomic Map