siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031764.3 | NP_113952.2 | 10 | 1415 | ||
| XM_006250226.2 | XP_006250288.1 | 11 | 4712 | |||
| XM_006250227.2 | XP_006250289.1 | 13 | 2047 | |||
| XM_006250228.2 | XP_006250290.1 | 12 | 1828 | |||
| XM_006250229.2 | XP_006250291.1 | 12 | 1776 | |||
| XM_008769761.1 | XP_008767983.1 | 12 | 1790 | |||
| XM_008769762.1 | XP_008767984.1 | 12 | 1722 | |||
| XM_008769763.1 | XP_008767985.1 | 12 | 2028 | |||
| GenBank mRNA | BC161961.1 | - | 10 | 1415 | 9027893 | |
| BQ207033.1 | - | 3 | 445 | |||

Ddr2
Genomic Map