siRNA Details
| Gene Symbol: Slit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022632.2 | NP_072154.2 | 25, 26 | 3238 | ||
| XM_008770210.1 | XP_008768432.1 | 22, 23 | 2373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slit2| Gene Symbol: Slit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022632.2 | NP_072154.2 | 25, 26 | 3238 | ||
| XM_008770210.1 | XP_008768432.1 | 22, 23 | 2373 | |||