siRNA Details
| Gene Symbol: Ptprq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022925.1 | NP_075214.1 | 11, 12 | 1888 | ||
| GenBank mRNA | AF063249.1 | - | 11, 12 | 1888 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprq| Gene Symbol: Ptprq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022925.1 | NP_075214.1 | 11, 12 | 1888 | ||
| GenBank mRNA | AF063249.1 | - | 11, 12 | 1888 | ||
Set Membership: |
|