siRNA Details
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108265.1 | NP_001101735.1 | 6 | 1074 | ||
GenBank mRNA | BC168968.1 | - | 6 | 1060 | 7323102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108265.1 | NP_001101735.1 | 6 | 1074 | ||
GenBank mRNA | BC168968.1 | - | 6 | 1060 | 7323102 |