siRNA Details
| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013182.1 | NP_001013200.1 | 2 | 135 | ||
| XM_006245994.2 | XP_006246056.1 | 2 | 549 | |||
| GenBank mRNA | BC091238.1 | - | 2 | 135 | 7323726 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hn1l| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013182.1 | NP_001013200.1 | 2 | 135 | ||
| XM_006245994.2 | XP_006246056.1 | 2 | 549 | |||
| GenBank mRNA | BC091238.1 | - | 2 | 135 | 7323726 | |