siRNA Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009701.1 | NP_001009701.1 | 1028 | |||
| GenBank mRNA | BC088404.1 | - | 1028 | 7108395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itfg3| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009701.1 | NP_001009701.1 | 1028 | |||
| GenBank mRNA | BC088404.1 | - | 1028 | 7108395 | ||