siRNA Details
| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100727.1 | NP_001094197.1 | 4 | 701 | ||
| XM_006246298.2 | XP_006246360.1 | 4 | 758 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rufy1| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100727.1 | NP_001094197.1 | 4 | 701 | ||
| XM_006246298.2 | XP_006246360.1 | 4 | 758 | |||