siRNA Details
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108279.1 | NP_001101749.1 | 2 | 240 | ||
XM_008767841.1 | XP_008766063.1 | 3 | 335 | |||
XM_008767842.1 | XP_008766064.1 | 3 | 625 | |||
XM_008767843.1 | XP_008766065.1 | 3 | 325 | |||
GenBank mRNA | FQ223808.1 | - | 3 | 296 | ||
FQ224714.1 | - | 3 | 296 |
